siRNA Details
| Gene Symbol: Kctd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001142731.1 | NP_001136203.1 | 5 | 2614 | ||
| NM_134112.5 | NP_598873.2 | 5 | 930 | |||
| GenBank mRNA | AK016623.1 | - | 4 | 975 | ||
| AY899808.1 | - | 5 | 930 | |||
| BC022615.1 | - | 3 | 598 | |||
| BC087623.1 | - | 5 | 829 | |||

Kctd1
Genomic Map