siRNA Details
| Gene Symbol: Btaf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080706.1 | NP_001074175.1 | 11 | 1512 | ||
| GenBank mRNA | AB287295.1 | - | 11 | 1237 | ||
| AK141888.1 | - | 5 | 1959 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Btaf1| Gene Symbol: Btaf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080706.1 | NP_001074175.1 | 11 | 1512 | ||
| GenBank mRNA | AB287295.1 | - | 11 | 1237 | ||
| AK141888.1 | - | 5 | 1959 | |||