siRNA Details
| Gene Symbol: Uqcc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001160356.1 | NP_001153828.1 | 2 | 546 | ||
| GenBank mRNA | AK131602.1 | - | 2 | 537 | ||
| AK131892.1 | - | 2 | 510 | |||
| AK132116.1 | - | 2 | 537 | |||
| AY941793.1 | - | 2 | 537 | |||
| BC029863.1 | - | 2 | 471 | 4460168 | ||

Uqcc3
Genomic Map