siRNA Details
| Gene Symbol: Lgr4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172671.2 | NP_766259.2 | 12, 13 | 1593 | ||
| GenBank mRNA | AK044357.1 | - | 11, 12 | 901 | ||
| AK052660.1 | - | 1, 2 | 602 | |||
| CN667965.1 | - | 3, 4 | 165 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lgr4| Gene Symbol: Lgr4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172671.2 | NP_766259.2 | 12, 13 | 1593 | ||
| GenBank mRNA | AK044357.1 | - | 11, 12 | 901 | ||
| AK052660.1 | - | 1, 2 | 602 | |||
| CN667965.1 | - | 3, 4 | 165 | |||