siRNA Details
| Gene Symbol: Padi2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017226.1 | NP_058922.1 | 5, 6 | 581 | ||
| XM_008764215.1 | XP_008762437.1 | 5, 6 | 619 | |||
| GenBank mRNA | J05022.1 | - | 5, 6 | 581 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Padi2| Gene Symbol: Padi2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017226.1 | NP_058922.1 | 5, 6 | 581 | ||
| XM_008764215.1 | XP_008762437.1 | 5, 6 | 619 | |||
| GenBank mRNA | J05022.1 | - | 5, 6 | 581 | ||