siRNA Details
| Gene Symbol: Eif2b3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001111277.1 | NP_001104747.1 | 7, 8 | 856 | ||
| NM_175135.4 | NP_780344.2 | 7, 8 | 856 | |||
| GenBank mRNA | AK145856.1 | - | 7, 8 | 833 | ||
| AK172173.1 | - | 6, 7 | 738 | |||
| BC115657.1 | - | 5, 6 | 530 | 40103205 | ||
| BC115658.1 | - | 5, 6 | 530 | 40103206 | ||

Eif2b3
Genomic Map