siRNA Details
| Gene Symbol: Fam50b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138746.2 | NP_620085.2 | 2 | 356 | ||
| GenBank mRNA | AK016195.1 | - | 2 | 267 | ||
| BC049659.1 | - | 2 | 269 | |||
| Y18506.1 | - | 2 | 257 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam50b| Gene Symbol: Fam50b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138746.2 | NP_620085.2 | 2 | 356 | ||
| GenBank mRNA | AK016195.1 | - | 2 | 267 | ||
| BC049659.1 | - | 2 | 269 | |||
| Y18506.1 | - | 2 | 257 | |||