siRNA Details
| Gene Symbol: Smim13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135577.2 | NP_001129049.1 | 2 | 748 | ||
| GenBank mRNA | AK082625.1 | - | 2 | 355 | ||
| BC024659.1 | - | 2 | 420 | |||
| BC066054.1 | - | 2 | 732 | |||
| BC094222.1 | - | 2 | 732 | |||

Smim13
Genomic Map