siRNA Details
| Gene Symbol: Slc5a10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033227.2 | NP_001028399.1 | 6, 7 | 701 | ||
| GenBank mRNA | AK144033.1 | - | 6, 7 | 702 | ||
| AK165337.1 | - | 5, 6 | 497 | |||
| BC141325.1 | - | 5, 6 | 494 | 9055937 | ||

Slc5a10
Genomic Map