siRNA Details
Gene Symbol: Hsd17b7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017235.3 | NP_058931.1 | 2 | 245 | ||
XM_006250215.2 | XP_006250277.1 | 2 | 268 | |||
XM_006250216.2 | XP_006250278.1 | 2 | 268 | |||
XM_006250217.2 | XP_006250279.1 | 2 | 268 | |||
GenBank mRNA | FM073997.1 | - | 2 | 238 | ||
FQ113345.1 | - | 2 | 245 | |||
U44803.1 | - | 2 | 190 |
