siRNA Details
Gene Symbol: Itga1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033228.3 | NP_001028400.2 | 29 | 5794 | ||
GenBank mRNA | AK033533.1 | - | 601 | |||
AK053377.1 | - | 4 | 2193 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Itga1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033228.3 | NP_001028400.2 | 29 | 5794 | ||
GenBank mRNA | AK033533.1 | - | 601 | |||
AK053377.1 | - | 4 | 2193 |