siRNA Details
| Gene Symbol: C7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001243837.1 | NP_001230766.1 | 7 | 793 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C7| Gene Symbol: C7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001243837.1 | NP_001230766.1 | 7 | 793 | ||