siRNA Details
| Gene Symbol: Nr3c2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001083906.1 | NP_001077375.1 | 9 | 3555 | ||
| GenBank mRNA | AK136580.1 | - | 6 | 1598 | ||
| BC133713.1 | - | 9 | 3555 | 40143429 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nr3c2| Gene Symbol: Nr3c2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001083906.1 | NP_001077375.1 | 9 | 3555 | ||
| GenBank mRNA | AK136580.1 | - | 6 | 1598 | ||
| BC133713.1 | - | 9 | 3555 | 40143429 | ||