siRNA Details
| Gene Symbol: V1ra8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053223.1 | NP_444453.1 | 1 | 575 | ||
| GenBank mRNA | BC138392.1 | - | 1 | 599 | 8861412 | |
| BC138394.1 | - | 1 | 599 | 8861414 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

V1ra8| Gene Symbol: V1ra8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053223.1 | NP_444453.1 | 1 | 575 | ||
| GenBank mRNA | BC138392.1 | - | 1 | 599 | 8861412 | |
| BC138394.1 | - | 1 | 599 | 8861414 | ||