siRNA Details
| Gene Symbol: Stk33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_054103.1 | NP_473444.1 | 11 | 1482 | ||
| GenBank mRNA | AK014819.1 | - | 11 | 1482 | ||
| AM056057.1 | - | 8 | 750 | |||
| BC117798.1 | - | 8 | 771 | |||
| BC117799.1 | - | 8 | 771 | |||
| BC127158.1 | - | 8 | 771 | |||

Stk33
Genomic Map