siRNA Details
| Gene Symbol: Cd8a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081110.2 | NP_001074579.1 | 5 | 1396 | ||
| NM_009857.1 | NP_033987.1 | 4 | 1365 | |||
| GenBank mRNA | AK017889.1 | - | 5 | 1220 | ||
| AK037467.1 | - | 4 | 1244 | |||
| AK088128.1 | - | 5 | 1276 | |||
| AK088414.1 | - | 4 | 1240 | |||
| AK088447.1 | - | 4 | 1781 | |||
| AK137937.1 | - | 5 | 1247 | |||
| BC030679.1 | - | 5 | 1258 | 1247019 | ||

Cd8a
Genomic Map