siRNA Details
| Gene Symbol: Cryge | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007777.3 | NP_031803.3 | 3 | 611 | ||
| GenBank mRNA | AK014301.1 | - | 3 | 598 | ||
| BC057012.1 | - | 582 | ||||
| BC078417.1 | - | 586 | 6824205 | |||
| K02584.1 | - | 565 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cryge| Gene Symbol: Cryge | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007777.3 | NP_031803.3 | 3 | 611 | ||
| GenBank mRNA | AK014301.1 | - | 3 | 598 | ||
| BC057012.1 | - | 582 | ||||
| BC078417.1 | - | 586 | 6824205 | |||
| K02584.1 | - | 565 | ||||