siRNA Details
| Gene Symbol: Cryge | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007777.3 | NP_031803.3 | 3 | 586 | ||
| GenBank mRNA | AK014301.1 | - | 3 | 573 | ||
| BC057012.1 | - | 557 | ||||
| BC078417.1 | - | 561 | 6824205 | |||
| BC132506.1 | - | 3 | 586 | 40130783 | ||
| BC132508.1 | - | 3 | 586 | 40130785 | ||
| K02584.1 | - | 540 | ||||

Cryge
Genomic Map