siRNA Details
| Gene Symbol: Ctla2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145801.1 | NP_001139273.1 | 2, 3 | 233 | ||
| NM_007797.1 | NP_031823.1 | 2, 3 | 233 | |||
| GenBank mRNA | AK150917.1 | - | 2, 3 | 192 | ||
| AY034578.1 | - | 2, 3 | 173 | |||
| BE448128.1 | - | 2, 3 | 233 | |||
| X15592.1 | - | 2, 3 | 173 | |||

Ctla2b
Genomic Map