siRNA Details
| Gene Symbol: Map2k5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033987.1 | NP_001029159.1 | 3, 4 | 844 | ||
| NM_017246.2 | NP_058942.1 | 3, 4 | 844 | |||
| XM_006243229.2 | XP_006243291.1 | 3, 4 | 903 | |||
| GenBank mRNA | BC078860.1 | - | 3, 4 | 844 | 7122708 | |
| U37462.1 | - | 3, 4 | 332 | |||
| U37464.1 | - | 3, 4 | 332 | |||

Map2k5
Genomic Map