siRNA Details
| Gene Symbol: Ddn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013741.1 | NP_001013763.1 | 2 | 2818 | ||
| GenBank mRNA | AK122363.1 | - | 2 | 2693 | ||
| AK158894.1 | - | 2 | 2819 | |||
| AK164154.1 | - | 1 | 2427 | |||
| BC157995.1 | - | 2 | 2564 | 9088145 | ||
| BC157996.1 | - | 2 | 2564 | 9088146 | ||

Ddn
Genomic Map