siRNA Details
| Gene Symbol: Scn10a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017247.1 | NP_058943.1 | 2 | 319 | ||
| XM_008766673.1 | XP_008764895.1 | 2 | 370 | |||
| GenBank mRNA | X92184.1 | - | 2 | 319 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Scn10a| Gene Symbol: Scn10a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017247.1 | NP_058943.1 | 2 | 319 | ||
| XM_008766673.1 | XP_008764895.1 | 2 | 370 | |||
| GenBank mRNA | X92184.1 | - | 2 | 319 | ||