siRNA Details
| Gene Symbol: Egfbp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010115.6 | NP_034245.3 | 4 | 559 | ||
| GenBank mRNA | AB016032.1 | - | 4 | 535 | ||
| X58628.1 | - | 4 | 559 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Egfbp2| Gene Symbol: Egfbp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010115.6 | NP_034245.3 | 4 | 559 | ||
| GenBank mRNA | AB016032.1 | - | 4 | 535 | ||
| X58628.1 | - | 4 | 559 | |||