siRNA Details
| Gene Symbol: Erbb4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010154.1 | NP_034284.1 | 3, 4 | 724 | ||
| GenBank mRNA | AK144050.1 | - | 3, 4 | 725 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Erbb4| Gene Symbol: Erbb4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010154.1 | NP_034284.1 | 3, 4 | 724 | ||
| GenBank mRNA | AK144050.1 | - | 3, 4 | 725 | ||