siRNA Details
| Gene Symbol: Fbrs | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010183.1 | NP_034313.1 | 11 | 924 | ||
| GenBank mRNA | AK037952.1 | - | 7 | 704 | ||
| BC027193.1 | - | 9 | 795 | |||
| CF532387.1 | - | 5 | 594 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fbrs| Gene Symbol: Fbrs | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010183.1 | NP_034313.1 | 11 | 924 | ||
| GenBank mRNA | AK037952.1 | - | 7 | 704 | ||
| BC027193.1 | - | 9 | 795 | |||
| CF532387.1 | - | 5 | 594 | |||