siRNA Details
| Gene Symbol: Gdf11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010272.1 | NP_034402.1 | 3 | 1666 | ||
| GenBank mRNA | KC561772.1 | - | 2778 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gdf11| Gene Symbol: Gdf11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010272.1 | NP_034402.1 | 3 | 1666 | ||
| GenBank mRNA | KC561772.1 | - | 2778 | |||