siRNA Details
| Gene Symbol: Gjb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017251.2 | NP_058947.1 | 2 | 450 | ||
| XM_006257061.2 | XP_006257123.1 | 3 | 602 | |||
| XM_008773255.1 | XP_008771477.1 | 2 | 1333 | |||
| GenBank mRNA | BC078868.1 | - | 2 | 414 | 7127952 | |
| CK476084.1 | - | 2 | 450 | |||
| FQ210131.1 | - | 2 | 435 | |||
| X04070.1 | - | 1 | 396 | |||

Gjb1
Genomic Map