siRNA Details
| Gene Symbol: Kif7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001291222.1 | NP_001278151.1 | 2 | 452 | ||
| NM_010626.3 | NP_034756.2 | 2 | 452 | |||
| GenBank mRNA | AB001435.1 | - | 2 | 157 | ||
| BC141045.1 | - | 2 | 453 | 9055657 | ||
| BC145222.1 | - | 2 | 453 | 9053773 | ||
| BU701630.1 | - | 1 | 100 | |||
| CD348088.1 | - | 1 | 57 | |||

Kif7
Genomic Map