siRNA Details
| Gene Symbol: Klra9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010651.3 | NP_034781.2 | 1 | 71 | ||
| GenBank mRNA | U49865.1 | - | 71 | |||
| U49866.1 | - | 71 | ||||
| U49867.1 | - | 71 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Klra9| Gene Symbol: Klra9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010651.3 | NP_034781.2 | 1 | 71 | ||
| GenBank mRNA | U49865.1 | - | 71 | |||
| U49866.1 | - | 71 | ||||
| U49867.1 | - | 71 | ||||