siRNA Details
| Gene Symbol: Lama3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010680.1 | NP_034810.1 | 35, 36 | 4568 | ||
| GenBank mRNA | X84014.1 | - | 16, 17 | 2286 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lama3| Gene Symbol: Lama3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010680.1 | NP_034810.1 | 35, 36 | 4568 | ||
| GenBank mRNA | X84014.1 | - | 16, 17 | 2286 | ||