siRNA Details
| Gene Symbol: Lama3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010680.1 | NP_034810.1 | 39, 40 | 5101 | ||
| GenBank mRNA | AK138740.1 | - | 2, 3 | 310 | ||
| X84013.1 | - | 2, 3 | 307 | |||
| X84014.1 | - | 20, 21 | 2819 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lama3| Gene Symbol: Lama3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010680.1 | NP_034810.1 | 39, 40 | 5101 | ||
| GenBank mRNA | AK138740.1 | - | 2, 3 | 310 | ||
| X84013.1 | - | 2, 3 | 307 | |||
| X84014.1 | - | 20, 21 | 2819 | |||