siRNA Details
Gene Symbol: Vmn1r9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134185.2 | NP_598946.2 | 1 | 50 | ||
GenBank mRNA | BC132241.1 | - | 1 | 50 | 40130518 | |
BC132577.1 | - | 1 | 50 | 40130854 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Vmn1r9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134185.2 | NP_598946.2 | 1 | 50 | ||
GenBank mRNA | BC132241.1 | - | 1 | 50 | 40130518 | |
BC132577.1 | - | 1 | 50 | 40130854 |