siRNA Details
| Gene Symbol: Vmn1r222 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134240.1 | NP_599001.1 | 1 | 136 | ||
| GenBank mRNA | BC120858.1 | - | 1 | 214 | 40129781 | |
| BC120860.1 | - | 1 | 214 | 40129783 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vmn1r222| Gene Symbol: Vmn1r222 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134240.1 | NP_599001.1 | 1 | 136 | ||
| GenBank mRNA | BC120858.1 | - | 1 | 214 | 40129781 | |
| BC120860.1 | - | 1 | 214 | 40129783 | ||