siRNA Details
Gene Symbol: Slc22a27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134256.1 | NP_599017.1 | 4 | 746 | ||
GenBank mRNA | AB056442.1 | - | 4 | 746 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Slc22a27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134256.1 | NP_599017.1 | 4 | 746 | ||
GenBank mRNA | AB056442.1 | - | 4 | 746 |