siRNA Details
| Gene Symbol: Pcsk5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163144.1 | NP_001156616.1 | 21 | 4237 | ||
| GenBank mRNA | AK032736.1 | - | 1 | 991 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pcsk5| Gene Symbol: Pcsk5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163144.1 | NP_001156616.1 | 21 | 4237 | ||
| GenBank mRNA | AK032736.1 | - | 1 | 991 | ||