siRNA Details
| Gene Symbol: Pcsk6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001291184.1 | NP_001278113.1 | 21 | 3201 | ||
| NM_011048.1 | NP_035178.1 | 22 | 3240 | |||
| GenBank mRNA | AK081039.1 | - | 4 | 758 | ||
| BC003302.1 | - | 6 | 967 | |||
| BC025946.1 | - | 6 | 971 | |||
| BC037450.1 | - | 19 | 2806 | |||
| BC058542.1 | - | 1 | 63 | |||

Pcsk6
Genomic Map