siRNA Details
| Gene Symbol: Scand1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020255.3 | NP_064651.2 | 2 | 405 | ||
| GenBank mRNA | AK009115.1 | - | 2 | 268 | ||
| CA451199.1 | - | 2 | 234 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Scand1| Gene Symbol: Scand1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020255.3 | NP_064651.2 | 2 | 405 | ||
| GenBank mRNA | AK009115.1 | - | 2 | 268 | ||
| CA451199.1 | - | 2 | 234 | |||