siRNA Details
| Gene Symbol: Fam195b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033231.2 | NP_001028403.1 | 5 | 1179 | ||
| GenBank mRNA | AK147780.1 | - | 5 | 1157 | ||
| AK210372.1 | - | 1 | 295 | |||
| BC002190.1 | - | 4 | 1111 | |||
| BC003940.1 | - | 4 | 1111 | |||
| BC025536.1 | - | 5 | 1152 | |||
| BC052229.1 | - | 4 | 1282 | |||

Fam195b
Genomic Map