siRNA Details
| Gene Symbol: Eme2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163102.1 | NP_001156574.1 | 8 | 1501 | ||
| GenBank mRNA | AK012738.1 | - | 7 | 1561 | ||
| BC034637.1 | - | 1540 | ||||
| BC092228.1 | - | 8 | 1459 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Eme2| Gene Symbol: Eme2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163102.1 | NP_001156574.1 | 8 | 1501 | ||
| GenBank mRNA | AK012738.1 | - | 7 | 1561 | ||
| BC034637.1 | - | 1540 | ||||
| BC092228.1 | - | 8 | 1459 | |||