siRNA Details
| Gene Symbol: Cnksr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081047.1 | NP_001074516.1 | 14 | 1278 | ||
| GenBank mRNA | AK145910.1 | - | 14 | 1240 | ||
| BC113805.1 | - | 14 | 1243 | 40062208 | ||
| BC113956.1 | - | 14 | 1243 | 40062209 | ||

Cnksr1
Genomic Map