siRNA Details
Gene Symbol: Tmprss11a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033233.2 | NP_001028405.1 | 9 | 1165 | ||
GenBank mRNA | AK142820.1 | - | 9 | 1166 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Tmprss11a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033233.2 | NP_001028405.1 | 9 | 1165 | ||
GenBank mRNA | AK142820.1 | - | 9 | 1166 |