siRNA Details
| Gene Symbol: Zzef1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001045536.2 | NP_001039001.1 | 31 | 5050 | ||
| GenBank mRNA | AK134006.1 | - | 10 | 1612 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zzef1| Gene Symbol: Zzef1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001045536.2 | NP_001039001.1 | 31 | 5050 | ||
| GenBank mRNA | AK134006.1 | - | 10 | 1612 | ||