siRNA Details
| Gene Symbol: Pramef6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001085414.2 | NP_001078883.2 | 5, 6 | 1476 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pramef6| Gene Symbol: Pramef6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001085414.2 | NP_001078883.2 | 5, 6 | 1476 | ||