siRNA Details
| Gene Symbol: Nhs | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081052.2 | NP_001074521.1 | 6 | 1730 | ||
| NM_001290526.1 | NP_001277455.1 | 7 | 1793 | |||
| GenBank mRNA | BC147632.1 | - | 6 | 1730 | 9056622 | |
| BC158016.1 | - | 7 | 1793 | 9007322 | ||
| BC172134.1 | - | 7 | 1793 | 9054804 | ||

Nhs
Genomic Map