siRNA Details
| Gene Symbol: Zc3h7b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081016.1 | NP_001074485.1 | 23 | 5796 | ||
| GenBank mRNA | AJ223207.1 | - | 1 | 82 | ||
| AK147484.1 | - | 23 | 5563 | |||
| AK171949.1 | - | 15 | 2588 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zc3h7b| Gene Symbol: Zc3h7b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081016.1 | NP_001074485.1 | 23 | 5796 | ||
| GenBank mRNA | AJ223207.1 | - | 1 | 82 | ||
| AK147484.1 | - | 23 | 5563 | |||
| AK171949.1 | - | 15 | 2588 | |||