siRNA Details
| Gene Symbol: Serpina1c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009245.2 | NP_033271.1 | 5 | 1222 | ||
| GenBank mRNA | BC009818.1 | - | 1165 | 4211464 | ||
| BC010984.1 | - | 1165 | 4163602 | |||
| BC010988.1 | - | 1163 | ||||
| BC011041.1 | - | 1166 | 4193647 | |||
| BC015266.1 | - | 1117 | ||||
| BC021325.1 | - | 1206 | 5052898 | |||
| BC021780.1 | - | 1185 | 4160260 | |||
| BC024108.1 | - | 1176 | 5099206 | |||
| BC031707.1 | - | 1152 | 5123976 | |||
| M75720.1 | - | 5 | 1159 | |||

Serpina1c
Genomic Map