siRNA Details
| Gene Symbol: Ccdc154 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001079929.2 | NP_001073398.2 | 16 | 2197 | ||
| GenBank mRNA | AK133084.1 | - | 17 | 2314 | ||
| BB017594.2 | - | 5 | 458 | |||
| JN935900.1 | - | 15 | 1816 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccdc154| Gene Symbol: Ccdc154 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001079929.2 | NP_001073398.2 | 16 | 2197 | ||
| GenBank mRNA | AK133084.1 | - | 17 | 2314 | ||
| BB017594.2 | - | 5 | 458 | |||
| JN935900.1 | - | 15 | 1816 | |||