siRNA Details
Gene Symbol: Ccdc154 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079929.2 | NP_001073398.2 | 8 | 1173 | ||
GenBank mRNA | AK133084.1 | - | 9 | 1270 | ||
JN935900.1 | - | 7 | 792 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ccdc154 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079929.2 | NP_001073398.2 | 8 | 1173 | ||
GenBank mRNA | AK133084.1 | - | 9 | 1270 | ||
JN935900.1 | - | 7 | 792 |