siRNA Details
| Gene Symbol: Hectd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144788.2 | NP_659037.2 | 19 | 3246 | ||
| GenBank mRNA | AK085720.1 | - | 11 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hectd1| Gene Symbol: Hectd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144788.2 | NP_659037.2 | 19 | 3246 | ||
| GenBank mRNA | AK085720.1 | - | 11 | |||