siRNA Details
| Gene Symbol: Chmp6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001085498.2 | NP_001078967.1 | 6, 7 | 558 | ||
| GenBank mRNA | AK162346.1 | - | 2, 3 | 135 | ||
| AK172123.1 | - | 5, 6 | 707 | |||
| BC147754.1 | - | 6, 7 | 553 | 9056703 | ||
| BC147755.1 | - | 6, 7 | 553 | 9056704 | ||
| BG078687.2 | - | 6, 7 | 473 | |||

Chmp6
Genomic Map